Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Veratrole-d2 | 203645-56-1 | 99.7% | 25 MG
SDP

Catalog No. 5000414387
Encompass_Preferred

Veratrole-d2 is a deuterium-labeled form of Veratrole (1,2-Dimethoxybenzene), a compound naturally found in plants. It plays a significant role in plant reproduction, species differentiation, and interactions with pollinators due to its circadian rhythm of release. Deuterium-labeled compounds like Veratrole-d2 are valuable in drug development as tracers for quantitation and for their potential to alter pharmacokinetic and metabolic profiles.

  • Deuterium labeled compound
  • Molecular formula C8H8D2O2
  • Molecular weight 140.18
  • High purity 99.7% (GC)
  • Isotopic enrichment 98.5%
  • Appearance is a colorless to light yellow liquid
  • Targets isotope-labeled compounds and cytochrome P450
  • Can be used as tracers for quantitation
  • Has potential to affect pharmacokinetic and metabolic profiles

Catalog No. 50-004-14387 Supplier Medchemexpress LLC Supplier No. 25MGHYB1812S1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Veratrole-d2 is a deuterium-labeled form of Veratrole (1,2-Dimethoxybenzene), a compound naturally found in plants. It plays a significant role in plant reproduction, species differentiation, and interactions with pollinators due to its circadian rhythm of release. Deuterium-labeled compounds like Veratrole-d2 are valuable in drug development as tracers for quantitation and for their potential to alter pharmacokinetic and metabolic profiles.

  • Deuterium labeled compound
  • Molecular formula C8H8D2O2
  • Molecular weight 140.18
  • High purity 99.7% (GC)
  • Isotopic enrichment 98.5%
  • Appearance is a colorless to light yellow liquid
  • Targets isotope-labeled compounds and cytochrome P450
  • Can be used as tracers for quantitation
  • Has potential to affect pharmacokinetic and metabolic profiles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.