Learn More
Medchemexpress LLC Veratrole-d2 | 203645-56-1 | 99.7% | 50 MG
SDP
Catalog No. 5000411412
Encompass_Preferred
Veratrole-d2 is a deuterium-labeled form of Veratrole (1,2-Dimethoxybenzene), a compound naturally found in plants that attracts pollinators. Its release exhibits a circadian rhythm, playing an important role in plant reproduction, species differentiation, and interactions with pollinators. This compound can also be demethylated by cytochrome P-450 in *Streptomyces setonii*. Deuterated compounds like Veratrole-d2 are utilized as tracers for quantification during drug development due to their potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled version of Veratrole
  • Key compound in plant reproduction and interactions with pollinators
  • Can be demethylated by cytochrome P-450
  • Used as a tracer for quantification in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-11412 Supplier Medchemexpress LLC Supplier No. HYB1812S150MG
Veratrole-d2 is a deuterium-labeled form of Veratrole (1,2-Dimethoxybenzene), a compound naturally found in plants that attracts pollinators. Its release exhibits a circadian rhythm, playing an important role in plant reproduction, species differentiation, and interactions with pollinators. This compound can also be demethylated by cytochrome P-450 in *Streptomyces setonii*. Deuterated compounds like Veratrole-d2 are utilized as tracers for quantification during drug development due to their potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled version of Veratrole
  • Key compound in plant reproduction and interactions with pollinators
  • Can be demethylated by cytochrome P-450
  • Used as a tracer for quantification in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs