Learn More
Medchemexpress LLC Veratrole-d4 (1,2-dimethoxybenzene-d4) | 126840-15-1 | 100 MG
SDP
Catalog No. 5000412736
Encompass_Preferred
Veratrole-d4 is a deuterium-labeled version of Veratrole (1,2-Dimethoxybenzene). Veratrole is a significant plant compound that widely exists in plants and attracts pollinators. Its release follows a circadian rhythm, impacting plant reproduction, species differentiation, and interactions with pollinators. Additionally, Veratrole can be demethylated by cytochrome P-450 in Streptomyces setonii.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12736 Supplier Medchemexpress LLC Supplier No. HYB1812S100MG
Veratrole-d4 is a deuterium-labeled version of Veratrole (1,2-Dimethoxybenzene). Veratrole is a significant plant compound that widely exists in plants and attracts pollinators. Its release follows a circadian rhythm, impacting plant reproduction, species differentiation, and interactions with pollinators. Additionally, Veratrole can be demethylated by cytochrome P-450 in Streptomyces setonii.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.