Learn More
Medchemexpress LLC Veratrole-d4 (1,2-Dimethoxybenzene-d4) | 126840-15-1 | 50 MG
SDP
Catalog No. 5000413052
Encompass_Preferred
Veratrole-d4 (1,2-Dimethoxybenzene-d4) is the deuterium-labeled form of Veratrole. Veratrole (1,2-Dimethoxybenzene) is a significant compound found in plants, where it plays a role in attracting pollinators, influencing plant reproduction, species differentiation, and interactions with pollinators. This compound also undergoes demethylation by cytochrome P-450 in Streptomyces setonii. Veratrole-d4 is intended for research use only.
  • Can be utilized as a tracer in research applications.
  • Suitable for use as an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes, which can potentially affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13052 Supplier Medchemexpress LLC Supplier No. HYB1812S50MG
Veratrole-d4 (1,2-Dimethoxybenzene-d4) is the deuterium-labeled form of Veratrole. Veratrole (1,2-Dimethoxybenzene) is a significant compound found in plants, where it plays a role in attracting pollinators, influencing plant reproduction, species differentiation, and interactions with pollinators. This compound also undergoes demethylation by cytochrome P-450 in Streptomyces setonii. Veratrole-d4 is intended for research use only.
  • Can be utilized as a tracer in research applications.
  • Suitable for use as an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes, which can potentially affect the pharmacokinetic and metabolic profiles of drugs.