Learn More
Medchemexpress LLC Veratrole-d6 | 24658-24-0 | 50 MG
SDP
Catalog No. 5000411175
Encompass_Preferred
Veratrole-d6 is a deuterium-labeled Veratrole (1,2-Dimethoxybenzene), a key compound found in plants that attracts pollinators. Its release shows a circadian rhythm, playing a vital role in plant reproduction, species differentiation, and interactions with pollinators. This compound can also be demethylated by cytochrome P-450 in *Streptomyces setonii*.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11175 Supplier Medchemexpress LLC Supplier No. HYB1812S350MG
Veratrole-d6 is a deuterium-labeled Veratrole (1,2-Dimethoxybenzene), a key compound found in plants that attracts pollinators. Its release shows a circadian rhythm, playing a vital role in plant reproduction, species differentiation, and interactions with pollinators. This compound can also be demethylated by cytochrome P-450 in *Streptomyces setonii*.
  • Stable heavy isotopes are incorporated into drug molecules for quantitation.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.