Learn More
Medchemexpress LLC Veratrole-d6 | 24658-24-0 | 98.7% | 100 MG
SDP
Catalog No. 5000412632
Encompass_Preferred
Veratrole-d6 is a deuterium-labeled version of Veratrole, also known as 1,2-Dimethoxybenzene. Veratrole is an important compound found in plants, playing a role in attracting pollinators. Its release follows a circadian rhythm and is crucial for plant reproduction, species differentiation, and interactions with pollinators. Additionally, Veratrole can undergo demethylation by cytochrome P-450 in *Streptomyces setonii*.
  • Isotope-labeled compound
  • Targets cytochrome P450
Catalog No. 50-004-12632 Supplier Medchemexpress LLC Supplier No. HYB1812S3100MG
Veratrole-d6 is a deuterium-labeled version of Veratrole, also known as 1,2-Dimethoxybenzene. Veratrole is an important compound found in plants, playing a role in attracting pollinators. Its release follows a circadian rhythm and is crucial for plant reproduction, species differentiation, and interactions with pollinators. Additionally, Veratrole can undergo demethylation by cytochrome P-450 in *Streptomyces setonii*.
  • Isotope-labeled compound
  • Targets cytochrome P450