Learn More
Selleck Chemical LLC Veratryl alcohol S3213-100mg
SDP
Catalog No. 503109105
Encompass_Preferred
Veratryl alcohol (VA Veratrole alcohol 3 4-Dimethoxybenzyl alcohol) a secondary metabolite of some lignin degrading fungi is the natural substrate of Lignin peroxidase (LiP)
Catalog No. 50-310-9105 Supplier Selleck Chemical LLC Supplier No. S3213100MG
Veratryl alcohol (VA Veratrole alcohol 3 4-Dimethoxybenzyl alcohol) a secondary metabolite of some lignin degrading fungi is the natural substrate of Lignin peroxidase (LiP)