Learn More
MSC Veratti 1000 Safety Glasses, Black, Light Silver ScratchCoat, 1 ea
Catalog No. 5000794392
Encompass
These safety glasses feature a European style, wrap-around design with a lightweight polycarbonate frame and a 10 base curve lens, providing 99.9% UVA & UVB protection. They are equipped with high-impact polycarbonate lenses and a ScratchCoat lens coating for enhanced durability, complying with ANSI Z87.1+ standards.
  • European style, wrap-around design
  • One piece lightweight polycarbonate frame
  • Three point adjustable temples
  • Features a 10 base curve lens
  • High impact polycarbonate lenses
  • ScratchCoat lens coating
  • Provides 99.9% UVA & UVB protection to 385Nm
  • Complies with ANSI Z87.1+
Catalog No. 50-007-94392 Supplier MSC Supplier No. 48072300
These safety glasses feature a European style, wrap-around design with a lightweight polycarbonate frame and a 10 base curve lens, providing 99.9% UVA & UVB protection. They are equipped with high-impact polycarbonate lenses and a ScratchCoat lens coating for enhanced durability, complying with ANSI Z87.1+ standards.
  • European style, wrap-around design
  • One piece lightweight polycarbonate frame
  • Three point adjustable temples
  • Features a 10 base curve lens
  • High impact polycarbonate lenses
  • ScratchCoat lens coating
  • Provides 99.9% UVA & UVB protection to 385Nm
  • Complies with ANSI Z87.1+