Learn More
MSC VERATTI READER SFTY GLS
Catalog No. 5000787356
Encompass
Encon Veratti GT Readers safety glasses feature integrated bifocal lenses, designed to enhance vision for close tasks. Their lightweight, low-profile design ensures comfort during prolonged wear and provides comprehensive eye protection.
  • Improves vision for close objects with integrated bifocal lenses
  • Weightless, low profile design ensures all-day comfort
  • Ultra flexible frame structure for durability
  • Rubberized temple tips for a secure fit
  • Features a 9.75 base curve lens for wrap-around protection
  • High impact polycarbonate lenses resist damage
  • ScratchCoat® coating provides enhanced scratch resistance
  • Offers 99.9% UVA & UVB protection to 385 Nm
  • Meets ANSI/ISEA Z87.1:2020 and CAN/CSA Z94.3:20 safety standards
Catalog No. 50-007-87356 Supplier MSC Supplier No. 59412411
Encon Veratti GT Readers safety glasses feature integrated bifocal lenses, designed to enhance vision for close tasks. Their lightweight, low-profile design ensures comfort during prolonged wear and provides comprehensive eye protection.
  • Improves vision for close objects with integrated bifocal lenses
  • Weightless, low profile design ensures all-day comfort
  • Ultra flexible frame structure for durability
  • Rubberized temple tips for a secure fit
  • Features a 9.75 base curve lens for wrap-around protection
  • High impact polycarbonate lenses resist damage
  • ScratchCoat® coating provides enhanced scratch resistance
  • Offers 99.9% UVA & UVB protection to 385 Nm
  • Meets ANSI/ISEA Z87.1:2020 and CAN/CSA Z94.3:20 safety standards