Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Verbascose, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014564 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4564 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4564 Supplier MilliporeSigma Supelco Supplier No. 562175MG
Add to Cart
Add to Cart

Verbascose, a pentasaccharide that contains four units of galactose and one unit of sucrose, occurs mostly in leguminous plants. In plants it is synthesized by an enzyme named, multifunctional stachyose synthase using raffinose as well as stachyose as a galactosyl acceptor.

TRUSTED_SUSTAINABILITY

Specifications

CAS 546-62-3
Physical Form Neat
Linear Formula C30H52O26
MDL Number MFCD03095487
Synonym alpha-D-Galp-(1->6)-alpha-D-Galp-(1->6)-alpha-D-Galp-(1->6)-alpha-D-Glup(1<->2)-beta-D-Fruf
Type Analytical Standard
Quantity 5 mg
Formula Weight 828.72
Percent Purity ≥97% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.