Learn More
Medchemexpress LLC HY-N0021 10mg Medchemexpress, Verbascoside CAS:61276-17-3 Purity:>98%
SDP
Catalog No. 501933359
Encompass_Preferred
Medchemexpress, HY-N0021 10mg Verbascoside CAS:61276-17-3 Verbascoside is isolated from Lantana camara, acts as an ATP-competitive inhibitor of PKC , with an IC 50 of 25 µM, and has antitumor, anti-inflammatory and antineuropathic pain activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3359 Supplier Medchemexpress LLC Supplier No. HYN002110MG
Medchemexpress, HY-N0021 10mg Verbascoside CAS:61276-17-3 Verbascoside is isolated from Lantana camara, acts as an ATP-competitive inhibitor of PKC , with an IC 50 of 25 µM, and has antitumor, anti-inflammatory and antineuropathic pain activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.