Learn More
Selleck Chemical LLC Verbascoside S5458-1mg
SDP
Catalog No. 503110689
Encompass_Preferred
Verbascoside (Acteoside Kusaginin) a phenylpropanoid glycoside from lemon verbena has several biological properties such as anti-inflammatory antimicrobial antitumor and antioxidant
Catalog No. 50-311-0689 Supplier Selleck Chemical LLC Supplier No. S54581MG
Verbascoside (Acteoside Kusaginin) a phenylpropanoid glycoside from lemon verbena has several biological properties such as anti-inflammatory antimicrobial antitumor and antioxidant