Learn More
TARGETMOL CHEMICALS INC VERBASCOSIDE 25MG
SDP
Catalog No. 502580292
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Verbascoside (Kusaginin) is a protein kinase C inhibitor with antimicrobial anti-inflammatory activities. purity: 97%
Catalog No. 50-258-0292 Supplier TARGETMOL CHEMICALS INC Supplier No. T337525MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Verbascoside (Kusaginin) is a protein kinase C inhibitor with antimicrobial anti-inflammatory activities. purity: 97%