Learn More
Enterprise Technology Solutions Verbatim SDHC Cards, 6 Hours, Speed Class 10, 16GB, 4/BX
SDP
Catalog No. 5000739693
Encompass_Preferred
Premium SDHC Card offers optimal performance for your SDHC digital camera or other SDHC slot-equipped device. Card features the industry's highest performance rating (Speed Class 6 rated), and minimum write speed of (60X) 9MB per second and read speed of (133X) 20MB per second. SDHC card delivers superior video capture/playback characteristics and fast photo upload/downloads to/from your PC. Card requires a SDHC host device. Write Protect Switch prevents accidental data deletion. SDHC card is fully compliant with all applicable SDHC standards.
Catalog No. 50-007-39693 Supplier Enterprise Technology Solutions Supplier No. VER96808BX
Premium SDHC Card offers optimal performance for your SDHC digital camera or other SDHC slot-equipped device. Card features the industry's highest performance rating (Speed Class 6 rated), and minimum write speed of (60X) 9MB per second and read speed of (133X) 20MB per second. SDHC card delivers superior video capture/playback characteristics and fast photo upload/downloads to/from your PC. Card requires a SDHC host device. Write Protect Switch prevents accidental data deletion. SDHC card is fully compliant with all applicable SDHC standards.