Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Verbenalin | 548-37-8 | 388.37 | 5 MG
SDP

Catalog No. 5000416418
Encompass_Preferred

Verbenalin is an orally active terpenoid glycoside extracted from the medicinal plant *Verbena officinalis*. It exhibits anti-inflammatory, antiviral, and neuroprotective effects. It binds strongly to the nsp-12 protein of the SARS-CoV-2 virus and is used in research for inflammatory and nervous system diseases such as hepatitis and Alzheimer's disease.

  • Orally active terpenoid glycoside
  • Exhibits anti-inflammatory effects
  • Exhibits antiviral effects
  • Exhibits neuroprotective effects
  • Binds strongly to nsp-12 protein of SARS-CoV-2 virus
  • Used in research for inflammatory diseases
  • Used in research for nervous system diseases
  • Purity: 99.90%
  • Appearance: Solid
  • Color: White to off-white
  • For research use only

Catalog No. 50-004-16418 Supplier Medchemexpress LLC Supplier No. HYN20145MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Verbenalin is an orally active terpenoid glycoside extracted from the medicinal plant *Verbena officinalis*. It exhibits anti-inflammatory, antiviral, and neuroprotective effects. It binds strongly to the nsp-12 protein of the SARS-CoV-2 virus and is used in research for inflammatory and nervous system diseases such as hepatitis and Alzheimer's disease.

  • Orally active terpenoid glycoside
  • Exhibits anti-inflammatory effects
  • Exhibits antiviral effects
  • Exhibits neuroprotective effects
  • Binds strongly to nsp-12 protein of SARS-CoV-2 virus
  • Used in research for inflammatory diseases
  • Used in research for nervous system diseases
  • Purity: 99.90%
  • Appearance: Solid
  • Color: White to off-white
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.