Learn More
Med Vet International Verbrugge Forceps 6.5", J0213VA
Catalog No. 5000585855
Encompass_Preferred
Verbrugge Bone Holding Forceps. Preferred instrument for holding bone plates or bone fragments. Self-centering with spinlock. Stainless steel. Bone Holding Forceps. Small. 150mm (6 1/2") for 2.7 and 3.5mm narrow plates.
Catalog No. 50-005-85855 Supplier Med Vet International Supplier No. JOR213VA
Verbrugge Bone Holding Forceps. Preferred instrument for holding bone plates or bone fragments. Self-centering with spinlock. Stainless steel. Bone Holding Forceps. Small. 150mm (6 1/2") for 2.7 and 3.5mm narrow plates.