Learn More
Med Vet International Verbrugge Forceps 9.5", J0213VB
Catalog No. 5000584411
Encompass_Preferred
Verbrugge Bone Holding Forceps. Preferred instrument for holding bone plates or bone fragments. Self-centering with spinlock. Stainless steel. Bone Holding Forceps. Large. 240mm (9 1/2") for 3.5 and 4.5mm plates.
Catalog No. 50-005-84411 Supplier Med Vet International Supplier No. JOR213VB
Verbrugge Bone Holding Forceps. Preferred instrument for holding bone plates or bone fragments. Self-centering with spinlock. Stainless steel. Bone Holding Forceps. Large. 240mm (9 1/2") for 3.5 and 4.5mm plates.