Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vercirnon sodium | 886214-18-2 | 98.0% | 466.91 g/mol | C22H20ClN2NaO4S | 10 MG
SDP

Catalog No. 5000287704
Encompass_Preferred

Vercirnon sodium is a selective, orally bioavailable antagonist of the chemokine receptor CCR9 supplied for research use. It is provided as a solid and as a DMSO solution for use in biochemical and cellular assays.

  • Selective CCR9 antagonist for research applications.
  • Available as solid forms and as a 10 mM solution in DMSO.
  • High purity (~98.0%) suitable for analytical and pharmacological studies.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Recommended storage: 4°C under inert gas; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-87704 Supplier Medchemexpress LLC Supplier No. HY15724A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vercirnon sodium is a selective, orally bioavailable antagonist of the chemokine receptor CCR9 supplied for research use. It is provided as a solid and as a DMSO solution for use in biochemical and cellular assays.

  • Selective CCR9 antagonist for research applications.
  • Available as solid forms and as a 10 mM solution in DMSO.
  • High purity (~98.0%) suitable for analytical and pharmacological studies.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Recommended storage: 4°C under inert gas; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.