Learn More
Selleck Chemical LLC Verdinexor
SDP
Catalog No. 5000004637
Encompass_Preferred
Verdinexor (KPT-335 ATG-527) is an orally bioavailable selective XPO1/CRM1 inhibitor
Catalog No. 50-000-04637 Supplier Selleck Chemical LLC Supplier No. S770725MG
Verdinexor (KPT-335 ATG-527) is an orally bioavailable selective XPO1/CRM1 inhibitor