Learn More
Selleck Chemical LLC Verdinexor (KPT-335) 5mg 1392136-43-4 ATG-527
SDP
Catalog No. 501365156
Encompass_Preferred
Verdinexor (KPT-335, ATG-527) is an orally bioavailable, selective XPO1/CRM1 inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5156 Supplier Selleck Chemical LLC Supplier No. S77075MG
Verdinexor (KPT-335, ATG-527) is an orally bioavailable, selective XPO1/CRM1 inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.