Learn More
Cayman Chemical Verdiperstat, Purity: ≥98%, CAS: 890655-80-8, 10 mg
SDP
Catalog No. 502915187
Encompass_Preferred
An irreversible inhibitor of MPO (IC50 = 630 nM); selective for MPO over a panel of 130 enzymes, transporters, and ion channels (IC50s = >50 µM for all); reduces MPO expression, prevents an increase in neuronal loss, and prevents decreases in motor score and stride length in a PLP-αSYN transgenic mouse model of multiple system atrophy at 180 µmol/kg, p.o.
Catalog No. 50-291-5187 Supplier Cayman Chemical Supplier No. 3733310 MG
An irreversible inhibitor of MPO (IC50 = 630 nM); selective for MPO over a panel of 130 enzymes, transporters, and ion channels (IC50s = >50 µM for all); reduces MPO expression, prevents an increase in neuronal loss, and prevents decreases in motor score and stride length in a PLP-αSYN transgenic mouse model of multiple system atrophy at 180 µmol/kg, p.o.