Learn More
Cayman Chemical VerdIperstat 50mg
SDP
Supplier:  Cayman Chemical 3733350 MG
Encompass_Preferred
An irreversible inhibitor of MPO (IC50 = 630 nM); selective for MPO over a panel of 130 enzymes, transporters, and ion channels (IC50s = >50 µM for all); reduces MPO expression, prevents an increase in neuronal loss, and prevents decreases in motor score and stride length in a PLP-αSYN transgenic mouse model of multiple system atrophy at 180 µmol/kg, p.o.
Catalog No. 50-291-5325