Learn More
Phenomenex Inc Verex Cap (pre-assembled), 10-425, w/
Catalog No. 502400239 Shop All Phenomenex Products
Encompass_Preferred
Verex Cap (pre-assembled), 10-425, w/ PTFE/Silicone preSlit septa, Black
Catalog No. 50-240-0239 Supplier Phenomenex Inc Supplier No. AR0807713
Verex Cap (pre-assembled), 10-425, w/ PTFE/Silicone preSlit septa, Black