Learn More
Phenomenex Inc Verex Cap (pre-assembled), 9mm, w/
Catalog No. 502397024 Shop All Phenomenex Products
Encompass_Preferred
Verex Cap (pre-assembled), 9mm, w/ Bonded-in PTFE/Silicone preSlit septa, blk
Catalog No. 50-239-7024 Supplier Phenomenex Inc Supplier No. AR0897713B
Verex Cap (pre-assembled), 9mm, w/ Bonded-in PTFE/Silicone preSlit septa, blk