Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 18 mm Screw, Round Bottom, Clear 33, No Patch

Supplier:  Phenomenex Inc AR0328012

Encompass_Preferred

Verex™ Headspace Vial, 23x75mm, 20mL AR0-3280-12

Catalog No. 50-239-8181


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.