Learn More
Phenomenex Inc Verex HS Screw Cap, 18mm, Magnetic, slv
Catalog No. 502400255 Shop All Phenomenex Products
Encompass_Preferred
Verex HS Screw Cap, 18mm, Magnetic, slv PTFE/Silicone septa (red/white)
Catalog No. 50-240-0255 Supplier Phenomenex Inc Supplier No. AR0815M12
Verex HS Screw Cap, 18mm, Magnetic, slv PTFE/Silicone septa (red/white)