Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 0.060in

Catalog No. 502397807 Shop All Phenomenex Products
Encompass_Preferred

Verex™ Septa, 8mm Dia., PTFE/Silicone AR0-6853-12

Catalog No. 50-239-7807 Supplier Phenomenex Inc Supplier No. AR0685312
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Verex™ Septa, 8mm Dia., PTFE/Silicone AR0-6853-12

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.