Learn More
Phenomenex Inc Verex Vial Kit, Snap Cap, 2mL
Catalog No. 502400677 Shop All Phenomenex Products
Encompass_Preferred
Verex Vial Kit, Snap Cap, 2mL Clear 33, w/ Patch + PTFE/Silicone
Catalog No. 50-240-0677 Supplier Phenomenex Inc Supplier No. AR0972112
Verex Vial Kit, Snap Cap, 2mL Clear 33, w/ Patch + PTFE/Silicone