Learn More
Bio Rad Laboratories VeriCheck ddPCR Empty-Full Capsid Kit, Discontinued, Assay Technology: Droplet Digital PCR (ddPCR), Measurements: Capsid Titer, Genome Titer, Empty-Full Ratio, Minimum Sample Input: 2 μl, AAV Serotypes Detected: AAV2, AAV5, AAV8, AAV9
Catalog No. NC3405024
Encompass
VeriCheck ddPCR Empty-Full Capsid Kit, Discontinued, Assay Technology: Droplet Digital PCR (ddPCR), Measurements: Capsid Titer, Genome Titer, Empty-Full Ratio, Minimum Sample Input: 2 μl, AAV Serotypes Detected: AAV2, AAV5, AAV8, AAV9
Catalog No. NC3405024 Supplier Bio Rad Laboratories Supplier No. 17010082
VeriCheck ddPCR Empty-Full Capsid Kit, Discontinued, Assay Technology: Droplet Digital PCR (ddPCR), Measurements: Capsid Titer, Genome Titer, Empty-Full Ratio, Minimum Sample Input: 2 μl, AAV Serotypes Detected: AAV2, AAV5, AAV8, AAV9