Learn More
Medchemexpress LLC Vericiguat-d3 hydrochloride | 1350653-20-1 | 99.8% | 465.86 | 1 MG
SDP
Catalog No. 5000393493
Encompass_Preferred
Vericiguat-d3 (BAY1021189-d3) hydrochloride is a deuterated Vericiguat with tracer functionality. Vericiguat is an orally available stimulator of guanylate cyclase.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-93493 Supplier Medchemexpress LLC Supplier No. HY16774S1MG
Vericiguat-d3 (BAY1021189-d3) hydrochloride is a deuterated Vericiguat with tracer functionality. Vericiguat is an orally available stimulator of guanylate cyclase.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.