Learn More
Medchemexpress LLC Vericiguat-d3 hydrochloride | 1350653-20-1 | 99.8% | 465.86 | C19H14D3ClF2N8O2 | 10 MG
SDP
Catalog No. 5000289336
Encompass_Preferred
Vericiguat-d3 hydrochloride is the deuterated hydrochloride salt of vericiguat supplied for analytical and research use. It is designed to provide tracer functionality and to serve as an internal standard in quantitative assays such as NMR, GC-MS, and LC-MS.
  • Provides deuterium-labeled tracer functionality for analytical studies.
  • Intended as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • High purity (≈99.8%) suitable for analytical applications.
  • Solid powder form; protect from light and store under nitrogen.
  • Recommended solvent storage: -80°C up to 6 months, -20°C up to 1 month.
Catalog No. 50-002-89336 Supplier Medchemexpress LLC Supplier No. HY16774S10MG
Vericiguat-d3 hydrochloride is the deuterated hydrochloride salt of vericiguat supplied for analytical and research use. It is designed to provide tracer functionality and to serve as an internal standard in quantitative assays such as NMR, GC-MS, and LC-MS.
  • Provides deuterium-labeled tracer functionality for analytical studies.
  • Intended as an internal standard for NMR, GC-MS, and LC-MS quantitation.
  • High purity (≈99.8%) suitable for analytical applications.
  • Solid powder form; protect from light and store under nitrogen.
  • Recommended solvent storage: -80°C up to 6 months, -20°C up to 1 month.