Learn More
Lovibond™ Verification Standard Kit MD100
Catalog No. 09051527 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric) Sample:
6 x 10 mL
Catalog No. Volume (Metric) Sample
09-051-527 6 x 10 mL
1 options
Catalog No. 09-051-527 Supplier Lovibond™ Supplier No. 215670
Add to Cart
Add to Cart
The kit allows you to confirm your device is taking accurate measurements.
  • The verification standard kit for the MD 100 / MD 200 is designed to assure the user of the accuracy and the reliability of the results related to the integrated wave lengths
  • The kit contains one zero standard, 6 different vials for checking 6 different wave lengths and allows checking the complete range of MD100 colorimeters
  • When stored and used as directed, the product is guaranteed to have a minimum remaining shelf life of 12 months.
Specifications
Type Verification Standard
Volume (Metric) Sample 6 x 10 mL