Learn More
Lovibond™ Verification Standard Kit MD600/ MD610
Catalog No. 09053297 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric) Sample:
6 x 10 mL
Catalog No. Volume (Metric) Sample
09-053-297 6 x 10 mL
1 options
Catalog No. 09-053-297 Supplier Lovibond™ Supplier No. 215640
Add to Cart
Add to Cart
The kit can be used to verify that all 6 wavelengths are measuring correctly.
  • Includes case, test certificate, zero vial, and verification standard vials: 430 nm, 530 nm, 560 nm, 580 nm, 610 nm, 660 nm
  • When stored and used as directed, the product is guaranteed to have a minimum remaining shelf life of 12 months.
Specifications
Type Verification Standard
Volume (Metric) Sample 6 x 10 mL