Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Cesium Ionization Buffer for AAS 2% Cs in 1% HNO3, Ricca Chemical
SDP

Catalog No. p-7073881 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010141500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010141500 Supplier Ricca Chemical Company Supplier No. RV010141500N

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural Poly Bottle
CAS 534-17-8, 7697-37-2, 7732-18-5
CAS Min % 94.15%, 2.87%, 0.98%
CAS Max % 97.99%, 2.99%, 1.02%
Specific Gravity 1.01
Physical Form Liquid
Molecular Formula CCs2O3
MDL Number MFCD00010957
Melting Point Approximately 0°C
InChI Key FJDQFPXHSGXQBY-UHFFFAOYSA-L
SMILES [Cs+].[Cs+].[O-]C([O-])=O
IUPAC Name dicaesium(1+) carbonate
Molecular Weight (g/mol) 325.82
Quantity 500 mL
PubChem CID 10796
Grade High Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.