Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Hydrogen Sulfite Standard for Ion Chromatography, 1000ppm in H2O, ISO 17025/Guide 34, Ricca Chemical
SDP

Catalog No. RV010951100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010951100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010951100 Supplier Ricca Chemical Company Supplier No. RV010951100C

missing translation for 'validMainframeMsg'

Formulated for high performance and reliable results

  • 1000ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7631-90-5 , 7732-18-5
Molecular Formula NaO3S
Molecular Weight (g/mol) 103.05
MDL Number MFCD00003530
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
PubChem CID 23665763
ChEBI CHEBI:26709
IUPAC Name sodium sulfite
SMILES [Na+].[O-]S([O-])=O

Specifications

Packaging Bottle, Poly Natural
CAS Min % 97.8726%, 0.1274%
CAS Max % 100%, 0.1326%
Specific Gravity 1.01
Physical Form Liquid
Quantity 100 mL
Grade High Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.