Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ ICP Calibration Standard 15, Ricca Chemical
SDP

Catalog No. RV010670100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010670100 100 mL
RV010670500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010670100 Supplier Ricca Chemical Company Supplier No. RV010670100N
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

500 ppm K; 50 ppm Al, AS, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn /nManufactured and Tested i

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Specifications

Packaging Natural LDPE Bottle
CAS Min % 4.90%, 0.07%, 0.02%, 0.01%, 0.01%
CAS Max % 5.10%, 0.07%, 0.02%, 0.01%, 0.01%
CAS 13548-38-4, 10325-94-7, 7778-39-4, 7697-37-2
Specific Gravity 1.07
Physical Form Liquid
UN Number UN3264
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Grade High Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.