Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Mercury (Hg) Standard for AAS 1000 ppm in 10% HNO3, Ricca Chemical
SDP

Catalog No. RV010024500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010024500 500 mL
RV010024100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. RV010024500 Supplier Ricca Chemical Company Supplier No. RV010024500C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • 1000 ppm in 10% HNO3
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 10045-94-0 , 7697-37-2 , 7732-18-5
Molecular Formula HgN2O6
Molecular Weight (g/mol) 324.60
MDL Number MFCD00011038
InChI Key ORMNPSYMZOGSSV-UHFFFAOYSA-N
PubChem CID 24864
IUPAC Name mercury(2+) dinitrate
SMILES [Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Amber Glass Bottle
CAS Min % 88.04%, 9.80%, 0.16%
CAS Max % 91.64%, 10.20%, 0.16%
Specific Gravity 1.02
Physical Form Liquid
UN Number UN2031
Melting Point Approximately 0°C
Quantity 500 mL
Grade High Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.