Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Niobium Standard for ICP/MS 100 ppm in 2% HNO3/HF, Ricca Chemical
SDP

Catalog No. RV010537100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010537100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RV010537100 Supplier Ricca Chemical Company Supplier No. RV010537100N
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 7783-68-8 , 7697-37-2 , 7732-18-5 , 7783-68-8
Molecular Formula F5Nb
Molecular Weight (g/mol) 187.90
MDL Number MFCD00011129
InChI Key AOLPZAHRYHXPLR-UHFFFAOYSA-I
IUPAC Name niobium(5+) pentafluoride
SMILES [F-].[F-].[F-].[F-].[F-].[Nb+5]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 95.53%, 1.96%, 0.49%, 0.02%
CAS Max % 99.43%, 2.04%, 0.51%, 0.02%
Specific Gravity 1
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade High Purity
Chemical Name or Material Niobium ICP-MS Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.