Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VeriSpec™ Triethanolamine - HN(C2H4OH)3+ Standard for Ion Chromatography 1000 ppm in H2O, Ricca Chemical
SDP

Catalog No. RV010948100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RV010948100 100 mL
RV010948250 250 mL
RV010948500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RV010948100 Supplier Ricca Chemical Company Supplier No. RV010948100N
Add to Cart
Add to Cart

Manufactured and Tested in an ISO 17025/Guide 34 Facility

  • 1000 ppm in H2O
  • Manufactured and tested in an ISO 17025/Guide 34 Accredited facility
  • Product accompanied with detailed Certificate of Analysis

Chemical Identifiers

CAS 102-71-6 , 7732-18-5
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
MDL Number MFCD00002855
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
PubChem CID 7618
ChEBI CHEBI:28621
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
SMILES OCCN(CCO)CCO

Specifications

Packaging Natural LDPE Bottle
CAS Min % 97.90%, 0.10%
CAS Max % 100%, 0.10%
Specific Gravity 1.01
Physical Form Liquid
Melting Point Approximately 0°C
Quantity 100 mL
Grade High Purity
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.