Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Veritor™ COVID Connectivity Expansion Bundle

Catalog No. B256143 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Detectable Analytes:
SARS-CoV-2 Antigen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
B256143 SARS-CoV-2 Antigen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B256143 Supplier BD Supplier No. 256143
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to support rapid detection testing.

  • Components included:
    • 3 x BD Veritor™ System for SARS-CoV-2 (CLIA-waived)
    • 1 x BD Veritor™ InfoWiFi Module
TRUSTED_SUSTAINABILITY

Specifications

CE Marker No
Clia Complexity Waived
Detectable Analytes SARS-CoV-2 Antigen
Type COVID Testing Kit
Disposable Single-use
Sterility Non-sterile
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.