Learn More
Description
- Components included:
- 2 x BD Veritor™ System for Rapid Detection of SARS-CoV-2 and Flu A+B
- 1 x BD Veritor™ System for Rapid Detection of RSV (CLIA-waived)
- 1 x BD Veritor™ System for Rapid Detection of Group A Strep (CLIA-waived)
- 1 x BD Veritor™ Plus Analyzer and BD Veritor™ InfoWiFi Module
Specifications
Specifications
| CE Marker | No |
| Clia Complexity | Waived (EUA) |
| Detectable Analytes | SARS-CoV-2 Nucleocapsid Antigen, Influenza A, Influenza B |
| Type | COVID Testing Kit |
| Disposable | Single-use |
| Form | Ready-to-Use (RTU) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.