Learn More
BD Veritor™ Plus Analyzer with Flu A+B CLIA Moderate Complex Flu Assay Kits (2 x 30)
Shop All BD Diagnostic Systems ProductsDescription
Veritor™ Plus Analyzer
This chromatographic instrument features advanced particle technology that enhances sensitivity and adaptive read technology to compensate for background and non-specific binding to reduce false-positive results. With Analyze Now and Walk-Away modes, the instrument provides lab-quality test results in just 10 minutes so you can make decisions while the patient is onsite.
- Cleared for use with nasal swabs and nasopharyngeal swabs
- Reduce hands on time and create a flexible workflow
- Improved traceability with InfoScan module
- Better connectivity with InfoSync module
- Number of reads per instrument: 3500
- Rechargeable lithium battery, print-enabled
Influenza A+B CLIA Moderate Complex Assay Kits
This system detects nucleoprotein antigens from nasopharyngeal wash, aspirate and swab in transport media samples from patients.
- Aids in rapid diagnosis of influenza A and B viral infections
- Tests samples processed from respiratory specimens
- Sensitive and specific for test to reduce false positives
- Components: Lateral flow test strips, RV reagent C in tubes (100μL per tube), 300μL pipettes, Flu A+/B- control swab, Flu B+/A- control swab
- Two kits included, sufficient for 60 assays (2 x 30)
Specifications
Specifications
| Description | Flu A + B Test Kit |
| Clia Complexity | Moderate |
| Detectable Analytes | Influenza A and B |
| Sample Type | Nasopharyngeal swabs, nasopharyngeal aspirate and nasopharyngeal washes |
| Type | Analyzer and Flu A+B Immunoassay Test Kit |
| For Use With (Application) | Detection of Flu A + B |
| Includes | Analyzer with lithium battery, two assay kits including: Lateral flow test strips, Pipettes, Flu A+/B- control swab, Flu B+/A- control swab, RV reagent C in tubes |
| Packaging Type | 1 Kit |
| Quantity | 2 x 30 Assays |
| Test Time | 10 min. |
For in vitro Diagnostic Use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.