Learn More
BD Veritor™ SARS‐CoV‐2 Control Swab Set
Catalog No. 21377302 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Detectable Analytes:
SARS-CoV-2
Catalog No. Detectable Analytes
21-377-302 SARS-CoV-2
1 options
Catalog No. 21-377-302 Supplier BD Supplier No. 256138
 Restricted Item
Add to Cart
Add to Cart
  • The BD Veritor™ SARS‐CoV‐2 Control Swab Set is a set of characterized positive and negative controls.
  • These controls are intended for use with the BD Veritor™ System for SARS‐CoV‐2 assay (Catalog Number 256092) as external quality control materials
TRUSTED_SUSTAINABILITY
Specifications
CE Marker No
Clia Complexity Waived
Detectable Analytes SARS-CoV-2
Type COVID Testing Kit Control
Disposable Single-use
Sterility Sterile
Form Ready-to-Use (RTU)