Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Veritor™ SARS-CoV-2 and Flu A+B Assay

Catalog No. B256097 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Veritor™ SARS-CoV-2 and Flu A+B Assay
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B256097 Veritor™ SARS-CoV-2 and Flu A+B Assay
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B256097 Supplier BD Supplier No. 256097

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Value pack contains 5 SARS-CoV-2 andFlu A+B kits

  • Rapidly detect and differentiate between SARS-CoV-2, Flu A, and Flu B from one sample on one test device
TRUSTED_SUSTAINABILITY

Specifications

CE Marker No
Clia Complexity Waived
Detectable Analytes Influenza A, Influenza B, SARS-CoV-2
Sample Type Anterior nasal specimen collection
Sensitivity 86.7% for SARS-CoV-2
Type Rapid antigen test
For Use With (Equipment) BD Veritor Plus Analyzer
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.