Learn More
BD Veritor™ System for SARS-CoV-2
Catalog No. 21377301 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Form:
Ready-to-Use (RTU)
Catalog No. Form
21-377-301 Ready-to-Use (RTU)
1 options
This item is not returnable. View return policy
Catalog No. 21-377-301 Supplier BD Supplier No. 256092
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Digital immunoassay for the rapid, detection of SARS‐CoV‐2 in anterior nasal swab specimens from individuals with signs and symptoms of upper respiratory infection
  • Displays easy-to-read digital results for COVID-19 in 15 minutes.
  • Provides clear, objective results by correcting for nonspecific binding and detecting positives not recognized by the unaided eye.
  • May reduce manual test processing errors with easy operations and single-button functionality.
  • Provides intuitive sample processing with prefilled, unitized reagent tubes. Adapts easily to your workflow by offering 2 operational modes.
TRUSTED_SUSTAINABILITY
Specifications
CE Marker No
Clia Complexity Waived
Detectable Analytes SARS-CoV-2
Type COVID Testing Kit
Disposable Single-use
Sterility Sterile
Form Ready-to-Use (RTU)