Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Veritor™ System SARS-CoV-2 and Flu A+B Control Swabs, 10 SARS+, 10 Flu A+, 10 Flu B+

Catalog No. 22029575 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
For Use With (Equipment):
Veritor™ System
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-029-575 Veritor™ System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-029-575 Supplier BD Supplier No. 256090
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The BD Veritor™ Plus System is a handheld device that enables testing for Flu A+B, Group A Strep, and respiratory syncytial virus (RSV) assays.

  • One-button functionality and workflow flexibility allow the healthcare team to confidently use the device.
  • In addition, the system's fast and accurate results may help improve the patient experience.
  • The BD Veritor™ Plus System streamlines the point-of-care testing experience.
TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes SARS-CoV-2, Flu A, Flu B
Format Kit
For Use With (Equipment) Veritor™ System
Sample Type Swab
Type Control Swab Kit
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.