Learn More
Uline Vermiculite Skid Lot - Grade 4
Catalog No. NC2096348
Encompass
This item must be ordered in multiples of 15 per supplier requirements
Vermiculite, 4 cubic foot bag for shipping liquids and chemicals, coarse grains, less dust, meets ICAA and IATA hazardous material requirements, 8mm particles, not for use with Hydrofluroic Acid
This item is not returnable. View return policy
Catalog No. NC2096348 Supplier Uline Supplier No. S3393S
This item must be ordered in multiples of 15 per supplier requirements
Vermiculite, 4 cubic foot bag for shipping liquids and chemicals, coarse grains, less dust, meets ICAA and IATA hazardous material requirements, 8mm particles, not for use with Hydrofluroic Acid