Learn More
Vermiculite
SCI_ED
Catalog No. S17729
Change view
Click to view available options
Quantity:
2 gal.
Catalog No. Quantity
S17729 2 gal.
1 options
Catalog No. S17729 Supplier Better Methods Supplier No. S17729
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
The choice for starting seedlings or cuttings.
  • Light, porous, inert mica medium
  • Use to lighten and aerate heavy soil
  • Packaged in 2 gal. container
Specifications
Product Type Vermiculite
For Use With (Application) Use to lighten and aerate heavy soil
Quantity 2 gal.