Learn More
MSC Vermont Gage Class X Go Plug Gage, 0.1567 in, 2 in
Catalog No. 5000515092
Encompass
This Class X Go Plug Gage is designed for high-precision measurement applications. Made of cold-stabilized tool steel, it features a 0.1567 in diameter and a 2 in length. It is ideal for calibrating measuring equipment or directly gaging parts, offering exceptional accuracy with included NIST traceability.
  • 0.1567 in diameter
  • 2 in length
  • Class X tolerance of +0.00004 in, -0.0000 in
  • Constructed from cold-stabilized tool steel, 60/65 Rc
  • Surface finish of 4 microinch or better
  • Suitable for calibrating measuring equipment or gaging parts directly
  • Includes a Certificate of Accuracy with NIST traceability
  • Round within 1/2 of the tolerance
Catalog No. 50-005-15092 Supplier MSC Supplier No. 91039743
This Class X Go Plug Gage is designed for high-precision measurement applications. Made of cold-stabilized tool steel, it features a 0.1567 in diameter and a 2 in length. It is ideal for calibrating measuring equipment or directly gaging parts, offering exceptional accuracy with included NIST traceability.
  • 0.1567 in diameter
  • 2 in length
  • Class X tolerance of +0.00004 in, -0.0000 in
  • Constructed from cold-stabilized tool steel, 60/65 Rc
  • Surface finish of 4 microinch or better
  • Suitable for calibrating measuring equipment or gaging parts directly
  • Includes a Certificate of Accuracy with NIST traceability
  • Round within 1/2 of the tolerance