Learn More
MSC Vermont Gage Class X Minus Pin Gage, 0.0320 in Diameter, 2 in Length
Catalog No. 5000516142
Encompass
This Class X Minus Pin Gage is designed for precision measurement tasks. It has a diameter of 0.0320 in and a length of 2 in, and features a 4 Micro Coating. This gage is suitable for the calibration of measuring equipment or for directly gaging parts.
  • Class X Minus tolerance grade
  • 0.0320 in diameter
  • 2 in length
  • 4 Micro Coated finish
  • Constructed from tool steel
  • Includes Certificate of Accuracy with NIST Traceability
  • Designed for calibration of measuring equipment or direct part gaging
Catalog No. 50-005-16142 Supplier MSC Supplier No. 88803200
This Class X Minus Pin Gage is designed for precision measurement tasks. It has a diameter of 0.0320 in and a length of 2 in, and features a 4 Micro Coating. This gage is suitable for the calibration of measuring equipment or for directly gaging parts.
  • Class X Minus tolerance grade
  • 0.0320 in diameter
  • 2 in length
  • 4 Micro Coated finish
  • Constructed from tool steel
  • Includes Certificate of Accuracy with NIST Traceability
  • Designed for calibration of measuring equipment or direct part gaging